Tisk…

Anti-HMMR Rabbit Monoclonal Antibody [clone: EPR4054]

Dodavatel: Abcam
ABCAAB124729-100EA 28830 CZK
ABCAAB124729-100
Anti-HMMR Rabbit Monoclonal Antibody [clone: EPR4054]
Protilátky
Type: Primary
Antigen: HMMR (hyaluronan-mediated motility receptor (RHAMM))
Clonality: monoclonal
Clone: EPR4054
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR