Tisk…

Anti-SKP2 Rabbit Monoclonal Antibody [clone: EPR3306]

Dodavatel: Abcam
ABCAAB124799-100EA 23870 CZK
ABCAAB124799-100
Anti-SKP2 Rabbit Monoclonal Antibody [clone: EPR3306]
Protilátky
Type: Primary
Antigen: SKP2 (S-phase kinase-associated protein 2, E3 ubiquitin protein ligase)
Clonality: monoclonal
Clone: EPR3306
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR