Tisk…
Anti-WNT5B Rabbit Monoclonal Antibody [clone: EPR7135]
Dodavatel: Abcam
ABCAAB124818-100EA 24700 CZK
ABCAAB124818-100
Anti-WNT5B Rabbit Monoclonal Antibody [clone: EPR7135]
Protilátky
Type: Primary
Antigen: WNT5B (wingless-type MMTV integration site family, member 5B)
Clonality: monoclonal
Clone: EPR7135
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Objednat