Tisk…

Anti-CDC25B Rabbit Monoclonal Antibody [clone: EPR3459]

Dodavatel: Abcam
ABCAAB124819-100EA 25460 CZK
ABCAAB124819-100
Anti-CDC25B Rabbit Monoclonal Antibody [clone: EPR3459]
Protilátky
Type: Primary
Antigen: CDC25B (cell division cycle 25B)
Clonality: monoclonal
Clone: EPR3459
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR