Tisk…

Anti-Nicotinic Acetylcholine Receptor alpha 4 Rabbit Monoclonal Antibody [clone: EPR4563]

Dodavatel: Abcam
ABCAAB124832-100EA 28250 CZK
ABCAAB124832-100
Anti-Nicotinic Acetylcholine Receptor alpha 4 Rabbit Monoclonal Antibody [clone: EPR4563]
Protilátky
Type: Primary
Antigen: Nicotinic Acetylcholine Receptor alpha 4
Clonality: monoclonal
Clone: EPR4563
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR