Tisk…
Anti-DOCK2 Rabbit Monoclonal Antibody [clone: EPR6647]
Dodavatel: Abcam
ABCAAB124838-100EA 25460 CZK
ABCAAB124838-100
Anti-DOCK2 Rabbit Monoclonal Antibody [clone: EPR6647]
Protilátky
Type: Primary
Antigen: DOCK2 (dedicator of cytokinesis 2)
Clonality: monoclonal
Clone: EPR6647
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Objednat