Tisk…

Anti-14-3-3 theta/tau Rabbit Monoclonal Antibody [clone: EPR5123]

Dodavatel: Abcam
ABCAAB124909-100EA 24120 CZK
ABCAAB124909-100
Anti-14-3-3 theta/tau Rabbit Monoclonal Antibody [clone: EPR5123]
Protilátky
Type: Primary
Antigen: 14-3-3 theta/tau (14-3-3 theta/tau)
Clonality: monoclonal
Clone: EPR5123
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR