Tisk…

Anti-PNLIP Rabbit Monoclonal Antibody [clone: EPR6276]

Dodavatel: Abcam
ABCAAB124915-100EA 24120 CZK
ABCAAB124915-100
Anti-PNLIP Rabbit Monoclonal Antibody [clone: EPR6276]
Protilátky
Type: Primary
Antigen: PNLIP (pancreatic lipase)
Clonality: monoclonal
Clone: EPR6276
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR