Tisk…
Anti-TLR7 Rabbit Monoclonal Antibody [clone: EPR2088]
Dodavatel: Abcam
ABCAAB124928-100EA 28310 CZK
ABCAAB124928-100
Anti-TLR7 Rabbit Monoclonal Antibody [clone: EPR2088]
Protilátky
Type: Primary
Antigen: TLR7 (toll-like receptor 7)
Clonality: monoclonal
Clone: EPR2088
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Objednat