Tisk…

Anti-TXNRD1 Rabbit Monoclonal Antibody [clone: EPNCIR129]

Dodavatel: Abcam
ABCAAB124954-100EA 26690 CZK
ABCAAB124954-100
Anti-TXNRD1 Rabbit Monoclonal Antibody [clone: EPNCIR129]
Protilátky
Type: Primary
Antigen: TXNRD1 (thioredoxin reductase 1)
Clonality: monoclonal
Clone: EPNCIR129
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR