Tisk…

Anti-IKBKB Rabbit Monoclonal Antibody [clone: EPR6043]

Dodavatel: Abcam
ABCAAB124957-100EA 28410 CZK
ABCAAB124957-100
Anti-IKBKB Rabbit Monoclonal Antibody [clone: EPR6043]
Protilátky
Type: Primary
Antigen: IKBKB (inhibitor of kappa light polypeptide gene enhancer in B-cells, kinase beta)
Clonality: monoclonal
Clone: EPR6043
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR