Tisk…

Anti-SRGAP2 Rabbit Monoclonal Antibody [clone: EP2506]

Dodavatel: Abcam
ABCAAB124958-100EA 23900 CZK
ABCAAB124958-100
Anti-SRGAP2 Rabbit Monoclonal Antibody [clone: EP2506]
Protilátky
Type: Primary
Antigen: SRGAP2 (SLIT-ROBO Rho GTPase activating protein 2)
Clonality: monoclonal
Clone: EP2506
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR