Tisk…

Anti-Cardiac FABP Rabbit monoclonal antibody unconjugated [clone: EPR6127]

Dodavatel: Abcam
ABCAAB124978-100EA 26540 CZK
ABCAAB124978-100
Anti-Cardiac FABP Rabbit monoclonal antibody unconjugated [clone: EPR6127]
Protilátky
Type: Primary
Antigen: Cardiac FABP
Clonality: monoclonal
Clone: EPR6127
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR