Tisk…

Anti-KAT7 Rabbit Monoclonal Antibody [clone: EPR7194]

Dodavatel: Abcam
ABCAAB124993-100EA 23900 CZK
ABCAAB124993-100
Anti-KAT7 Rabbit Monoclonal Antibody [clone: EPR7194]
Protilátky
Type: Primary
Antigen: KAT7 (K(lysine) acetyltransferase 7)
Clonality: monoclonal
Clone: EPR7194
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR