Tisk…
Anti-GSR Rabbit Monoclonal Antibody [clone: EPR7238]
Dodavatel: Abcam
ABCAAB124995-100EA 28870 CZK
ABCAAB124995-100
Anti-GSR Rabbit Monoclonal Antibody [clone: EPR7238]
Protilátky
Type: Primary
Antigen: GSR (glutathione reductase)
Clonality: monoclonal
Clone: EPR7238
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Objednat