Tisk…

Anti-Prorenin Rabbit Monoclonal Antibody [clone: EPR6433]

Dodavatel: Abcam
ABCAAB125012-100EA 26170 CZK
ABCAAB125012-100
Anti-Prorenin Rabbit Monoclonal Antibody [clone: EPR6433]
Protilátky
Type: Primary
Antigen: Prorenin
Clonality: monoclonal
Clone: EPR6433
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR