Tisk…

Anti-MYBBP1A Rabbit Monoclonal Antibody [clone: EPR7204]

Dodavatel: Abcam
ABCAAB125026-100EA 26020 CZK
ABCAAB125026-100
Anti-MYBBP1A Rabbit Monoclonal Antibody [clone: EPR7204]
Protilátky
Type: Primary
Antigen: MYBBP1A (MYB binding protein (P160) 1a)
Clonality: monoclonal
Clone: EPR7204
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR