Tisk…

Anti-EFEMP2 Rabbit monoclonal antibody unconjugated [clone: EPR684]

Dodavatel: Abcam
ABCAAB125073-100EA 23780 CZK
ABCAAB125073-100
Anti-EFEMP2 Rabbit monoclonal antibody unconjugated [clone: EPR684]
Protilátky
Type: Primary
Antigen: EFEMP2 (EGF containing fibulin-like extracellular matrix protein 2)
Clonality: monoclonal
Clone: EPR684
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR