Tisk…
Anti-SCAP Rabbit Polyclonal Antibody
Dodavatel: Abcam
ABCAAB125186-100EA 25800 CZK
ABCAAB125186-100
Anti-SCAP Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: SCAP (SREBF chaperone)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Objednat