Tisk…

Anti-GRB10 Rabbit Polyclonal Antibody

Dodavatel: Abcam
ABCAAB125583-100EA 26110 CZK
ABCAAB125583-100
Anti-GRB10 Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: GRB10 (growth factor receptor-bound protein 10)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR