Tisk…
Anti-GLUL Mouse Monoclonal Antibody [clone: 6/Glutamine Synthetase]
Dodavatel: Abcam
ABCAAB125724-100EA 25740 CZK
ABCAAB125724-100
Anti-GLUL Mouse Monoclonal Antibody [clone: 6/Glutamine Synthetase]
Protilátky
Type: Primary
Antigen: GLUL (glutamate-ammonia ligase)
Clonality: monoclonal
Clone: 6/Glutamine Synthetase
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human