Tisk…

Anti-IgG Goat Polyclonal Antibody (PE (Phycoerythrin))

Dodavatel: Abcam
ABCAAB125852-250EA 12050 CZK
ABCAAB125852-250
Anti-IgG Goat Polyclonal Antibody (PE (Phycoerythrin))
Protilátky
Type: Secondary
Antigen: Rhesus monkey IgG
Clonality: polyclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Goat
Isotype: IgG
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR