Tisk…

Anti-GIMAP6 Rabbit Polyclonal Antibody

Dodavatel: Abcam
ABCAAB126067-50EA 24420 CZK
ABCAAB126067-50
Anti-GIMAP6 Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: GIMAP6 (GTPase, IMAP family member 6)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR