Tisk…
Anti-PTPD2 Rabbit Polyclonal Antibody
Dodavatel: Abcam
ABCAAB126114-50EA 23380 CZK
ABCAAB126114-50
Anti-PTPD2 Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: PTPD2
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human