Tisk…

Anti-KIR2DS4 Rabbit Polyclonal Antibody

Dodavatel: Abcam
ABCAAB126128-50EA 24420 CZK
ABCAAB126128-50
Anti-KIR2DS4 Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: KIR2DS4 (killer cell immunoglobulin-like receptor, two domains, short cytoplasmic tail, 4)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR