Tisk…

Anti-GCSF Receptor Rabbit polyclonal antibody unconjugated

Dodavatel: Abcam
ABCAAB126167-50EA 26480 CZK
ABCAAB126167-50
Anti-GCSF Receptor Rabbit polyclonal antibody unconjugated
Protilátky
Type: Primary
Antigen: GCSF Receptor
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR