Tisk…

Anti-NSF Rabbit Polyclonal Antibody

Dodavatel: Abcam
AB126202-50
ABCAAB126202-50EA 27030 CZK
ABCAAB126202-50
Anti-NSF Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: NSF (N-ethylmaleimide-sensitive factor)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR