Tisk…
Anti-ABL2 Rabbit Polyclonal Antibody
Dodavatel: Abcam
ABCAAB126256-50EA 25740 CZK
ABCAAB126256-50
Anti-ABL2 Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: ABL2 (c-abl oncogene 2, non-receptor tyrosine kinase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human