Tisk…

Anti-NR1D1 Rabbit Polyclonal Antibody

Dodavatel: Abcam
ABCAAB126271-50EA 25740 CZK
ABCAAB126271-50
Anti-NR1D1 Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: NR1D1 (nuclear receptor subfamily 1, group D, member 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR