Tisk…
Anti-NRN1L Rabbit Polyclonal Antibody
Dodavatel: Abcam
ABCAAB126303-100EA 25010 CZK
ABCAAB126303-100
Anti-NRN1L Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: NRN1L (neuritin 1-like)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human