Tisk…
Anti-DCAF8L1 Rabbit polyclonal antibody unconjugated
Dodavatel: Abcam
ABCAAB126359-100EA 25010 CZK
ABCAAB126359-100
Anti-DCAF8L1 Rabbit polyclonal antibody unconjugated
Protilátky
Type: Primary
Antigen: DCAF8L1 (DDB1 and CUL4 associated factor 8-like 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human