Tisk…

Anti-SLC25A41 Rabbit Polyclonal Antibody

Dodavatel: Abcam
ABCAAB126372-100EA 25010 CZK
ABCAAB126372-100
Anti-SLC25A41 Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: SLC25A41 (solute carrier family 25, member 41)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR