Tisk…

Anti-SUPT5H Rabbit Monoclonal Antibody [clone: EPR5145]

Dodavatel: Abcam
ABCAAB126592-100EA 23810 CZK
ABCAAB126592-100
Anti-SUPT5H Rabbit Monoclonal Antibody [clone: EPR5145]
Protilátky
Type: Primary
Antigen: SUPT5H (suppressor of Ty 5 homolog (S. cerevisiae))
Clonality: monoclonal
Clone: EPR5145
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR