Tisk…

Anti-FTO Rabbit Monoclonal Antibody [clone: EPR6894]

Dodavatel: Abcam
ABCAAB126605-100EA 28620 CZK
ABCAAB126605-100
Anti-FTO Rabbit Monoclonal Antibody [clone: EPR6894]
Protilátky
Type: Primary
Antigen: FTO (fat mass and obesity associated)
Clonality: monoclonal
Clone: EPR6894
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR