Tisk…

Anti-SUMO4 Rabbit Monoclonal Antibody [clone: EPR7163]

Dodavatel: Abcam
ABCAAB126606-100EA 23900 CZK
ABCAAB126606-100
Anti-SUMO4 Rabbit Monoclonal Antibody [clone: EPR7163]
Protilátky
Type: Primary
Antigen: SUMO4 (small ubiquitin-like modifier 4)
Clonality: monoclonal
Clone: EPR7163
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR