Tisk…
Anti-HN1 Rabbit Monoclonal Antibody [clone: EPR7364]
Dodavatel: Abcam
ABCAAB126705-100EA 23350 CZK
ABCAAB126705-100
Anti-HN1 Rabbit Monoclonal Antibody [clone: EPR7364]
Protilátky
Type: Primary
Antigen: HN1 (hematological and neurological expressed 1)
Clonality: monoclonal
Clone: EPR7364
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Objednat