Tisk…

Anti-PPWD1 Rabbit Monoclonal Antibody [clone: EPR7440]

Dodavatel: Abcam
ABCAAB126710-100EA 23230 CZK
ABCAAB126710-100
Anti-PPWD1 Rabbit Monoclonal Antibody [clone: EPR7440]
Protilátky
Type: Primary
Antigen: PPWD1 (peptidylprolyl isomerase domain and WD repeat containing 1)
Clonality: monoclonal
Clone: EPR7440
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR