Tisk…
Anti-FMO3 Rabbit monoclonal antibody unconjugated [clone: EPR6968]
Dodavatel: Abcam
ABCAAB126711-100EA 27210 CZK
ABCAAB126711-100
Anti-FMO3 Rabbit monoclonal antibody unconjugated [clone: EPR6968]
Protilátky
Type: Primary
Antigen: FMO3 (flavin containing monooxygenase 3)
Clonality: monoclonal
Clone: EPR6968
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Objednat