Tisk…
Anti-STIP1 Rabbit Monoclonal Antibody [clone: EPR6605]
Dodavatel: Abcam
ABCAAB126724-100EA 26900 CZK
ABCAAB126724-100
Anti-STIP1 Rabbit Monoclonal Antibody [clone: EPR6605]
Protilátky
Type: Primary
Antigen: STIP1 (stress-induced phosphoprotein 1)
Clonality: monoclonal
Clone: EPR6605
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Objednat