Tisk…

Anti-STIP1 Rabbit Monoclonal Antibody [clone: EPR6606]

Dodavatel: Abcam
ABCAAB126753-100EA 26230 CZK
ABCAAB126753-100
Anti-STIP1 Rabbit Monoclonal Antibody [clone: EPR6606]
Protilátky
Type: Primary
Antigen: STIP1 (stress-induced phosphoprotein 1)
Clonality: monoclonal
Clone: EPR6606
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR