Tisk…

Anti-ARPC1B Rabbit Polyclonal Antibody

Dodavatel: Abcam
ABCAAB126941-50EA 25560 CZK
ABCAAB126941-50
Anti-ARPC1B Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: ARPC1B (actin related protein 2/3 complex, subunit 1B, 41kDa)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR