Tisk…
Anti-CCDC7 Rabbit Polyclonal Antibody
Dodavatel: Abcam
ABCAAB126989-50EA 23380 CZK
ABCAAB126989-50
Anti-CCDC7 Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: CCDC7 (coiled-coil domain containing 7)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human