Tisk…
Anti-GORASP2 Rabbit polyclonal antibody unconjugated
Dodavatel: Abcam
ABCAAB127024-50EA 25740 CZK
ABCAAB127024-50
Anti-GORASP2 Rabbit polyclonal antibody unconjugated
Protilátky
Type: Primary
Antigen: GORASP2 (golgi reassembly stacking protein 2, 55kDa)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human