Tisk…
Anti-TRIP4 Rabbit Polyclonal Antibody
Dodavatel: Abcam
ABCAAB127537-50EA 25740 CZK
ABCAAB127537-50
Anti-TRIP4 Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: TRIP4 (thyroid hormone receptor interactor 4)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human