Tisk…
Anti-LRRC1 Rabbit Polyclonal Antibody
Dodavatel: Abcam
ABCAAB127568-50EA 24420 CZK
ABCAAB127568-50
Anti-LRRC1 Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: LRRC1 (leucine rich repeat containing 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human