Tisk…
Anti-TRMT10A Rabbit Polyclonal Antibody
Dodavatel: Abcam
ABCAAB127687-50EA 24420 CZK
ABCAAB127687-50
Anti-TRMT10A Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: TRMT10A (tRNA methyltransferase 10 homolog A (S. cerevisiae))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human