Tisk…
Anti-ARHGEF25 Rabbit Polyclonal Antibody
Dodavatel: Abcam
ABCAAB127690-50EA 24420 CZK
ABCAAB127690-50
Anti-ARHGEF25 Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: ARHGEF25 (Rho guanine nucleotide exchange factor (GEF) 25)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human