Tisk…

Anti-RAD3 Rabbit Polyclonal Antibody

Dodavatel: Abcam
ABCAAB127891-50EA 24420 CZK
ABCAAB127891-50
Anti-RAD3 Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: RAD3
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR