Tisk…

Anti-GNAQ Goat polyclonal antibody unconjugated

Dodavatel: Abcam
ABCAAB128060-50EA 25740 CZK
ABCAAB128060-50
Anti-GNAQ Goat polyclonal antibody unconjugated
Protilátky
Type: Primary
Antigen: GNAQ (guanine nucleotide binding protein (G protein), q polypeptide)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR