Tisk…

Anti-PADI4 / PAD4 Mouse Monoclonal Antibody [clone: 4H5]

Dodavatel: Abcam
ABCAAB128086-100EA 26940 CZK
ABCAAB128086-100
Anti-PADI4 / PAD4 Mouse Monoclonal Antibody [clone: 4H5]
Protilátky
Type: Primary
Antigen: PADI4 / PAD4
Clonality: monoclonal
Clone: 4H5
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR